Learn More
HYDRANAL™ - LipoSolver CM, Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium), Honeywell Fluka™
Medium for volumetric one-component Karl Fischer titration in non-polar substances, fats and oils (chloroform-methanol-based)
Brand: Honeywell-Fluka 37855-6X1L
View more versions of this product
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
HYDRANAL™ - LipoSolver CM | |
∼1.25 g/mL | |
6 x 1 L | |
<90.0000%, <30.0000% | |
54°C | |
MFCD02687786 | |
Methylidyne trichloride |
Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium) | |
26°C | |
≥70.0000%, ≥20.0000% | |
67-56-1 | |
Glass Bottle | |
UN1992 | |
Karl Fischer |
Safety and Handling
- HYDRANAL-LipoSolver CM
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302 + H312-Harmful if swallowed or in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, methanol