missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Medium K, Honeywell Fluka™

Product Code. p-7122063 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15604360

Brand: Honeywell Fluka 346981L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free).

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

 

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Medium K
Boiling Point 58 °C
CAS Min % ≥30.00, ≥10.00
CAS Max % <30.00, <60.00
CAS 107-98-2, 75-89-8, 101023-55-6, 67-66-3
Density 1.25g/mL
Physical Form Liquid
Quantity 1 L
Flash Point 56°C (132.8°F)
Purity Grade Notes One component volumetry
Packaging Polycoated bottle
UN Number UN1992
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Medium K
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1A
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trichloromethane; chloroform, 1-methoxy-2-propanol; monopropylene glycol methylether, 2,2,2-Trifluo
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.