missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™

Product Code. 15664390 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15664390

missing translation for 'mfr': Honeywell Fluka 347414X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Methanol dry
Density 0.79g/mL
Quantity 4 x 2.5 L
Flash Point 9°C (48.2°F)
UN Number UN1230
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Methanol dry
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.