missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Methanol Rapid, Reagent for accelerated volumetric one-component KF titration (working medium), Honeywell Fluka™
Medium for accelerated volumetric one-component Karl Fischer titration.
Brand: Honeywell Fluka 37817-4X2.5L
View more versions of this product
This item is not returnable.
View return policy
Description
Specifications
HYDRANAL™-Methanol Rapid | |
11°C (51.8°F) | |
UN1230 |
0.80g/mL | |
4 x 2.5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Methanol Rapid
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, sulphur dioxide, imidazole
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction