missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - NEXTGEN Coulomat A-FA, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)
Brand: Honeywell Fluka 34771-500ML
This item is not returnable.
View return policy
Description
- HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
- Comprehensive product portfolio with 60 available products
- 40 years of innovation – check our NEXTGEN reagents HERE
- Dedicated technical service and support
- More than 800 Application Notes available on our website
- View our NEXTGEN Coulomat A-FA standard operating procedure
Specifications
HYDRANAL™ - NEXTGEN Coulomat A-FA | |
0.906 g/cm3 | |
500 mL | |
≥10.00, ≥2.00, ≥60.00, ≥2.00 | |
Glass Bottle | |
≤20.00, <10.00, <10.00, <70.00 | |
Soluble in water |
Orange | |
12°C to 13°C | |
5.5 | |
Confidential Stabilizer 2, 75-75-2, Confidential Stabilizer 1, 75-05-8 | |
Liquid | |
UN2924 |