missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titrations, for cells with and without diaphragm
£116.03 - £582.27
Specifications
Density | 0.893 g/cm3 (at 20°C) |
---|---|
Flash Point | 14°C |
Chemical Name or Material | HYDRANAL™ - NEXTGEN Coulomat AG-FI |
Color | Yellow |
CAS | 67-56-1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822370
|
Honeywell Fluka
34433-500ML |
500 mL |
£116.03
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16832370
|
Honeywell Fluka
34433-6X500ML |
6 x 500 mL |
£582.27
3 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Methanol-based
- Free of Imidazole
- Free of CMR Substances
Specifications
0.893 g/cm3 (at 20°C) | |
HYDRANAL™ - NEXTGEN Coulomat AG-FI | |
67-56-1 | |
64°C to 65°C (Methanol) | |
-98°C (Methanol) | |
UN1230 |
14°C | |
Yellow | |
≥60% | |
Glass Bottle | |
Liquid | |
Completely Miscible in Water |
Safety and Handling
Recommended Storage : Room Temperature
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL NEXTGEN Coulomat AG-FI
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, 2-ethylimidazole, 2-butylaminoethanol, sulphur dioxide, 2-ethyl imidazole hydroiodide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Honeywell Fluka™