missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Coulomat C-FA, Honeywell Fluka™

Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances).

Brand:  Honeywell Fluka 34470-6X50ML


 View more versions of this product

Product Code. 17594946

  • £1318.51 / 6 bottles

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
    • Comprehensive product portfolio with 60 available products
    • 40 years of innovation – check our NEXTGEN reagents HERE
    • Dedicated technical service and support
    • More than 800 Application Notes available on our website
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - NEXTGEN Coulomat C-FA
0.906 g/cm3
6 x 50 mL
5.5
<70.00, ≤20.00, <10.00, <10.00
Liquid
Soluble in water
Orange
12°C to 13°C
75-05-8, Confidential Stabilizer 1, 75-75-2, Confidential Stabilizer 2
≥2.00, ≥60.00, ≥2.00, ≥10.00
Ampoule
UN2924
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL NEXTGEN Coulomat C-FA

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : acetonitrile, methanesulphonic acid, 1-Ethylimidazole

SDS
Documents

Documents

Certificates
Special Offers

Special Offers