Learn More
HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Honeywell Fluka™
Brand: Honeywell Fluka 34812-6X1L
Code : NEW
Additional Details : CAS Number : 67-66-3 Weight : 7.56000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Specifications
100°C (212°F) | |
HYDRANAL™-Solvent CM | |
MFCD00147198 |
6 x 1 L | |
1.26g/mL | |
UN2810 |
Safety and Handling
- HYDRANAL-Solvent CM
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302 + H312-Harmful if swallowed or in contact with skin.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, methanol,imidazole, sulphur dioxide