missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solvent E, Honeywell Fluka™

Medium for volumetric two-component Karl Fischer titration (ethanol-based)

Brand:  Honeywell Fluka 34730-4X2.5L

Code : NEW

Additional Details : CAS Number : 288-32-4 Weight : 8.95000kg Transport : UN number : 1170 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15614370

  • £472.14 / Pack of 4
Estimated Shipment: 03-09-2024
to see stock.

Description

Description

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

Specifications

Specifications

12°C (53.6°F)
HYDRANAL™-Solvent E
64-17-5,111-42-2,288-32-4,7446-09-5
UN1170
4 x 2.5 L
0.90g/mL
MFCD01867183
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL Solvent E

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2,2'-iminodiethanol; diethanolamine,imidazole, sulphurdioxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers