missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Titrant 2, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™
Brand: Honeywell-Hydranal 34811-500ML
View more versions of this product
This item is not returnable.
View return policy
Description
Specifications
HYDRANAL™-Titrant 2 | |
11°C (51.8°F) | |
UN1230 |
0.80g/mL | |
500 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL Titrant 2
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methano; iodine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction