missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™

Product Code. 15654370 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
Unit Size:
1 L
100mL
2.5 L
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15654370

Brand: Honeywell-Hydranal 34732100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titrating agent for volumetric two-component Karl Fischer titration (ethanol-based)

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Titrant 5 E
Density 0.89g/mL
Quantity 100 mL
Flash Point 12°C (53.6°F)
UN Number UN1170
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.