missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Water-in-methanol 5.0, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Flash Point | 9°C (48.2°F) |
Quantity | 1 L |
UN Number | UN1230 |
Beilstein | 3662861 |
Formula Weight | 32.04g/mol |
Chemical Name or Material | HYDRANAL™-Water-in-methanol 5.0 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'