missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 x 4 mL Ampules
Specifications
Specifications
Density | 0.86g/mL at 20°C |
Flash Point | 25°C (77°F) |
Quantity | 10 x 4 mL Ampules |
UN Number | UN1307 |
Chemical Name or Material | HYDRANAL™-Water Standard 0.1 |
Safety and Handling



Product Identifier
- HYDRANAL
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : o-xylene
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction