missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 0.1 PC, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell Fluka™

Product Code. 15665825
Click to view available options
Quantity:
10 x 4 mL
Packaging:
Ampoule

Specifications

Density 1.205 g/cm3
Flash Point 135°C
Quantity 10 x 4 mL
CAS 108-32-7
CAS Max % ≤100.0000%
Boiling Point 242°C
Packaging Ampoule
Molecular Formula C4H6O3
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILES CC1COC(=O)O1
IUPAC Name 4-methyl-1,3-dioxolan-2-one
Molecular Weight (g/mol) 102.09
PubChem CID 7924
Grade Karl Fischer
Chemical Name or Material HYDRANAL™ - Water Standard 0.1 PC, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), exact value on report of analysis
Show More Show Less
compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.