missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
£60.70 - £316.89
Chemical Identifiers
CAS | 100-66-3, 108-32-7 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00007941 |
InChI Key | RDOXTESZEPMUJZ-UHFFFAOYSA-N |
Synonym | Methoxybenzene, Methyl phenyl ether |
PubChem CID | 7519 |
ChEBI | CHEBI:16579 |
IUPAC Name | anisole |
SMILES | COC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15664490
|
Honeywell-Hydranal
34828-6X40ML |
1 Kit |
£316.89
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15628380
|
Honeywell-Hydranal
34828-40ML |
40 mL |
£60.70
40mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
100-66-3, 108-32-7 | |
108.14 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
7519 | |
anisole |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
CHEBI:16579 | |
COC1=CC=CC=C1 |
Specifications
1.000 g/cm3 | |
100-66-3, 108-32-7 | |
<100.0000%, <10.0000% | |
Ampoule | |
HCONH2 | |
UN2222 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
anisole | |
7519 | |
Karl Fischer |
43°C | |
≥90.0000%, ≥1.0000% | |
153°C to 155°C | |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
COC1=CC=CC=C1 | |
108.14 | |
CHEBI:16579 | |
HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), exact value on report of analysis |
Safety and Handling
missing translation for 'shelfLife' : 1800 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Anisole solution
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL Water Standard 1.3
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Anisole, propylene carbonate