missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Water Standard KF-Oven 220°C-230°C, Standard for Karl Fischer oven check (water content ∼5.55%), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 g
6 x 10 g
Description
Advantages of HYDRANAL-Water Standards:
- Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
- Supplied with detailed instruction for use

Specifications
Specifications
Quantity | 10 g |
Packaging | Glass bottle |
UN Number | NONH for all modes of transport |
Chemical Name or Material | HYDRANAL™-Water Standard KF-Oven 220°C-230°C |
Safety and Handling
Product Identifier
- HYDRANAL Water Standard KF-Oven 220°C-230°C
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
-
Multiple Manufacturers
Chemicals Monthly Promotion
Take advantage of our exclusive monthly promotions on high-quality chemicals.Promo Code: 24198Expires: 2025-03-31
New Offer Save Up to 59%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HYDRANAL™ - Water Standard KF-Oven 220°C-230°C, Standard for Karl Fischer oven check (water content ∼5.55%), Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction