missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Working Medium K, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™

£94.10 - £315.47

Specifications

CAS 67-66-3, 107-07-3
MDL Number MFCD00147202
UN Number UN3382
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15644480
View Documents Promotion Details
Honeywell-Hydranal
34817-1L
1 L
£94.10
1 L
Please to purchase this item. Need a web account? Register with us today!
15654480
View Documents
Honeywell-Hydranal
34817-4X1L
4 x 1 L
£315.47
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

67-66-3, 107-07-3
MFCD00147202
UN3382
Videos
Safety and Handling

Safety and Handling

P260-P280-P302 + P352 + P310-P304 + P340 + P310-P305 + P351 + P338 + P310

Product Identifier
  • HYDRANAL-Working Medium K

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : trichloromethane; chloroform, 2-chloroethanol; ethylene chlorohydrin

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.