missing translation for 'onlineSavingsMsg'
Learn More

Whirl-pak™ Hydrated PolySponge™ Sampling Bag with Sampling Sponge
GREENER_CHOICE

Makes environmental surface sampling faster and easier

Brand:  Whirl-pak™ B01590WA

189.01 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 16688462

  • £239.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Specially designed for the Food & Beverage industry
  • Write-on label makes identifying samples easy and convenient
  • Bags are pre-hydrated with HiCap™ Neutralizing Broth
  • HiCap Broth provides superior neutralization, validated up to 72 hr.
  • Polyurethane sponges included with each bag are non toxic and do not contain biocides
  • Each box contains five foil pouches with 20 sponges each
  • Terminally gamma processed to guarantee sterility
  • Medical-grade, super absorbent, and ideal for rough surfaces
  • Compatible with 3M Petrifilm
  • 2 year shelf life
Specifications

Specifications

Sample Bag with Sampling Sponge
18 oz.
Environmental Monitoring
Sterile
16 mL
20/Pk.
Foil Pouch
2.50 mil
23 cm
11.5 cm
Without Glove
532 mL
Polyurethane
0.064 mm
100/Cs
Write-On Whirl-Pak™ Bag
Terminally Gamma Processed
9 in.
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers