missing translation for 'onlineSavingsMsg'
Learn More

Neogen Hydrated-Sponge

Product Code. p-7230040
Encompass
Click to view available options
Includes:
Sponge
Sponge, Gloves
Saturation Solution:
10 mL Buffered Peptone Water Broth
10 mL Letheen Broth
10 mL Neutralizing Buffer
Unit Size:
1 set
100 sets
This item is not returnable. View return policy

Product Code. 17298510

Brand: Neogen 7000002888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Uses simplified packaging to minimize handling and preparation, and reduces waste.

  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection.
  • Durable sponge withstands scrubbing.
  • Sponge bag with write-on label for ease in labeling samples.
  • Includes two decontaminated gloves for safety and convenience.

Specifications

Product Type Hydrated Sponge
Length (English) 3 in.
Width (English) 1.5 in.
Packaging Quantity 100/Cs.
Saturation Solution 10 mL Neutralizing Buffer
Includes Sponge, Gloves
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.