Learn More
Juchheim Laborgeraete™ Hydraulic Laboratory Jack
A laboratory lifting table with smooth-running lifting hydraulics that loads up to 85 kg.
£4675.00 - £6365.00
Specifications
Material Steel
View More Specs
Products 2
Product Code Brand Height (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Price Quantity & Availability  
10294772
View Documents
Juchheim Laborgeraete™
02294
625 mm
£4675.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10204782
View Documents
Juchheim Laborgeraete™
02296
1060 mm
£6365.00
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
  • For static and dynamic loads
  • Particularly stable due to scissor construction made of robust steel
  • Acid and base resistant due to high-quality powder coating
  • Requires no external connections (pump hydraulics fully integrated)
  • With handles for easy transport
  • To lower the load, the lever only has to be pressed slightly upwards. The load then sinks slowly (approx. 3 cm/s) and stops as soon as the lever is released again.
Specifications
Specifications
Steel
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Juchheim Laborgeraete™ Hydraulic Laboratory Jack