missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine acetate, 97%, Thermo Scientific Chemicals

Product Code. 10361301
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7335-65-1
Molecular Formula C2H8N2O2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00013141
InChI Key YFHNDHXQDJQEEE-UHFFFAOYSA-N
Synonym hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph
PubChem CID 165591
IUPAC Name acetic acid;hydrazine
SMILES NN.CC(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NH2NH2·CH3COOH
Quantity 25 g
Formula Weight 92.1
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Hydrazine acetate, 99%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'