missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine dihydrochloride, 98+%

Product Code. 11332988
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11332988

missing translation for 'mfr': Thermo Scientific Alfa Aesar 032725.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As Chlorine Scavenger in Hcl Gas Streams | CAS: 5341-61-7 | ClH5N2 | 68.50 g/mol

Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrazine;dihydrochloride
SMILES [H+].[Cl-].NN

Specifications

Boiling Point 200°C (decomposition)
Melting Point 198°C
Color White
Physical Form Lump
Quantity 100 g
Assay Percent Range >98%
Solubility Information Soluble in water. Slightly soluble in alcohol
Formula Weight 104.97
Odor Odorless
Appearance White crystalline or lump
Density 1.42g/cm3
Chemical Name or Material Hydrazine dihydrochloride, Special handling precautions required
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.