Learn More
Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 198.0°C |
Color | White |
Physical Form | Chunks and/or Powder |
Quantity | 100 g |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | NH2NH2·2HCl |
Merck Index | 15, 4809 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in alcohol |
Formula Weight | 104.97 |
Percent Purity | 99% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462797 | Certificate of Analysis | 15/05/2024 | 411765000,411761000,411760025 |
A0451036 | Certificate of Analysis | 02/05/2023 | 411765000,411761000,411760025 |
A0398533 | Certificate of Analysis | 14/12/2022 | 411765000,411761000,411760025 |
A0409467 | Certificate of Analysis | 13/12/2022 | 411765000,411761000,411760025 |
A0408785 | Certificate of Analysis | 13/12/2022 | 411765000,411761000,411760025 |
Safety and Handling



- Hydrazine dihydrochloride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Skin sensitiser Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrazine, dihydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.