missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine hydrate, 100% (Hydrazine, 64%), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 g |
Linear Formula | H2NNH2·XH2O |
Fieser | 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174 |
Merck Index | 14, 4771 |
Formula Weight | 32.04 |
Percent Purity | 100.00 to 100.93% (N2H4.xH2O), 64.00 to 64.60% (N2H4) |
Packaging | Glass bottle |
Refractive Index | 1.4285 to 1.4315 (20°C, 589nm) |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466638 | Certificate of Analysis | 08/10/2024 | 196715000,196711000 |
A0452817 | Certificate of Analysis | 16/05/2023 | 196715000,196711000 |
A0403947 | Certificate of Analysis | 14/12/2022 | 196715000,196711000 |
A0395151 | Certificate of Analysis | 14/12/2022 | 196715000,196711000 |
A0408836 | Certificate of Analysis | 13/12/2022 | 196715000,196711000 |
Safety and Handling




Product Identifier
- Hydrazine hydrate, 100% (Hydrazine, 64%)
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrazine hydrate, 100% (Hydrazine, 64%), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction