missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Hydrate, 99+%, Extra Pure, SLR, Fisher Chemical™
£419.00
Chemical Identifiers
CAS | 7803-57-8 |
---|---|
Molecular Formula | H6N2O |
Molecular Weight (g/mol) | 50.06 |
MDL Number | MFCD00149931 |
InChI Key | IKDUDTNKRLTJSI-UHFFFAOYSA-N |
Synonym | hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739Show More |
PubChem CID | 24654 |
ChEBI | CHEBI:35511 |
IUPAC Name | hydrazine;hydrate |
SMILES | O.NN |
Description
Chemical Identifiers
7803-57-8 | |
50.06 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
24654 | |
hydrazine;hydrate |
H6N2O | |
MFCD00149931 | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739Show More | |
CHEBI:35511 | |
O.NN |
Specifications
118°C | |
7803-57-8 | |
12 | |
H6N2O | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
O.NN | |
50.06 | |
CHEBI:35511 |
-51.7°C/-61.1°F | |
Colorless | |
Liquid | |
MFCD00149931 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
hydrazine;hydrate | |
24654 | |
50.06g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrazine hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H350-May cause cancer.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrazine Hydrate, 99+%, Extra Pure, SLR, Fisher Chemical™