missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Hydrate Solution, 20 to 24%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 L |
Linear Formula | NH2NH2 · H2O |
UN Number | UN3293 |
Beilstein | 878137 |
IUPAC Name | hydrazine;hydrate |
Formula Weight | 50.06g/mol |
Packaging | Poly bottle |
Appearance | Colorless |
Density | 1.01g/mL at 20°C |
Chemical Name or Material | Hydrazine hydrate solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'