missing translation for 'onlineSavingsMsg'
Learn More
Hydrazine monohydrate, 98+%, Thermo Scientific Chemicals Product Code.: 11447866

Hydrazine monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11447866
500 g
Click to view available options
Quantity:
50 g
250 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11447866

Brand: Thermo Scientific Alfa Aesar A14005.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

Solubility
Miscible with water and alcohol. Immiscible with chloroform, hydrocarbons and ether.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents, oxygen, copper, organic materials and zinc.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739Show MoreShow Less
PubChem CID 24654
ChEBI CHEBI:35511
SMILES O.NN

Specifications

Boiling Point 120°C to 121°C
Melting Point -52°C
Quantity 500 g
Linear Formula H2NNH2·H2O
UN Number UN2030
Flash Point 75°C (167°F)
Merck Index 14,4771
Solubility Information Miscible with water and alcohol. Immiscible with chloroform,hydrocarbons and ether.
IUPAC Name hydrazine;hydrate
Formula Weight 50.06 (32.05 Anhydrous)
Percent Purity ≥98%
Odor Amine-like
Refractive Index 1.431
Density 1.027
Chemical Name or Material Hydrazine monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253590 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog NumberA14005.0E,A14005.36,A14005.18,A14005.30
Lot Number10252718 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog NumberA14005.0E,A14005.36,A14005.18,A14005.30
Lot Number10251326 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14005.0E,A14005.36,A14005.18,A14005.30
Lot Number10249975 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA14005.0E,A14005.36,A14005.18,A14005.30
Lot Number10249362 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog NumberA14005.0E,A14005.36,A14005.18,A14005.30
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrazine monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrazine monohydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.