Learn More
Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals
Description
Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states. It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 120°C to 121°C |
Melting Point | -52°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 g |
Assay Percent Range | >99% |
Linear Formula | H2NNH2·H2O |
UN Number | 2030 |
Flash Point | 75° (167°F) |
Merck Index | 14,4771 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z05K013 | Certificate of Analysis | 06/12/2024 | 016651.36,016651.57,016651.A3,016651.22 |
U16K003 | Certificate of Analysis | 28/08/2024 | 016651.36,016651.57,016651.A3,016651.22 |
U29J026 | Certificate of Analysis | 21/09/2023 | 016651.36,016651.57,016651.A3,016651.22 |
N07I005 | Certificate of Analysis | 12/04/2022 | 016651.36,016651.57,016651.A3,016651.22 |
B21Y027 | Certificate of Analysis | 08/12/2021 | 016651.36,016651.57,016651.A3,016651.22 |
Safety and Handling




- Hydrazine monohydrate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H350-May cause cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrazine monohydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.