missing translation for 'onlineSavingsMsg'
Learn More
Hydrazine sulfate, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 15411627

Hydrazine sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 15411627
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 15411627

Brand: Thermo Scientific Alfa Aesar 040120.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Reagent for determination of C-terminal amino acidsHydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point ∽255°C (decomposition)
CAS 10034-93-2
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0% min
Molecular Formula H6N2O4S
MDL Number MFCD00044873
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
Solubility Information Soluble in water. Insoluble in alcohol.
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES NN.OS(=O)(=O)O
IUPAC Name hydrazine;sulfuric acid
Molecular Weight (g/mol) 130.118
PubChem CID 24842
Formula Weight 130.12
Grade ACS Reagent
Density 1.37 g/mL
Chemical Name or Material Hydrazine sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY29C047 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number040120.36
Lot NumberW27K059 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number040120.36
Lot NumberW27K061 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number040120.36
Lot NumberQ09K037 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number040120.36
Lot NumberY27J022 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number040120.36
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine sulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrazine sulphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrazine sulfate, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.