Learn More
Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar L10410.22
Code : NEW
Additional Details : CAS Number : 10034-85-2 Weight : 0.10000kg Transport : UN number : 1787 Chem class : 8 Pack group : II
Description
Hydriodic acid is used to convert primary alcohol into alkyl iodide. It is used in organic and inorganic synthesis as one of the primary sources of iodine as well as a reducing agent. It is also used to cleave the ethers to get alkyl iodides and alcohols.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hydriodic acid | |
10034-85-2,7732-18-5,6303-21-5 | |
1.701 g/mL | |
Plastic | |
HI |
-50.8°C | |
126°C | |
1.5 | |
100 g | |
127.91 |
Safety and Handling
- Hydriodic acid, 57%, stabilized with 1.5% hypophosphorous acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Hydriodic acid, hypo
RUO – Research Use Only