missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydriodic Acid, 64-68%, Certified AR for Analysis, Fisher Chemical
861.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 mL
Packaging:
Amber glass bottle
Specifications
pH | 1 |
Color | Brown-Red |
CAS | 10034-85-2 |
Melting Point | -51°C |
Boiling Point | 127°C |
Packaging | Amber glass bottle |
Quantity | 250 mL |
Molecular Formula | HI |
Physical Form | Liquid |
MDL Number | MFCD00011347 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Hydriodic acid, aqueous solution
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydriodic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'