Learn More
Hydriodic acid, ACS, 47%, stab. with 1.5% hypophosphorous acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Hydriodic acid is used as a catalyst in the manufacture of acetic acid, as a pharmaceutical intermediate, in various disinfectants and in the preparation of organic and inorganic iodides. It is also used to prepare alkyl iodides from primary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 mL |