missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydriodic acid, ACS, 55-58%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
Packaging:
4 x 500mL
Description
In organic chemistry, it is used as catalyst for the conversion of primary alcohol to alkyl halide. Suitable for use in methoxyl determinations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used as catalyst for conversion of primary alcohol to alkyl halide
- Suitable for use in methoxyl determinations.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Brown-Red to Yellow |
CAS | 10034-85-2 |
Density | 1.701 g/cm³ (14.195lb./gal) |
Heavy Metals (as Pb) | 0.001% |
Packaging | 4 x 500mL |
Residue after Ignition | 0.01% |
Sensitivity | Light sensitive liquid |
Vapor Pressure | 7100 hPa (5325 mm Hg) |
Quantity | 500 mL |
Molecular Formula | HI |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'