missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic Acid, no stabilizer, Honeywell

Product Code. 15641910
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
PubChem CID 24841
ChEBI CHEBI:43451
IUPAC Name hydrogen iodide
SMILES I

Specifications

Density 1.701g/mL at 25°C
Quantity 250 mL
UN Number 1787
Formula Weight 127.91g/mol
Percent Purity 99.99% (trace metals basis)
Chemical Name or Material Hydriodic Acid, Distilled, Contains No Stabilizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.