missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic Acid, Puriss p.a., ≥67% (T), Honeywell™

Product Code. 15695870
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
PubChem CID 24841
ChEBI CHEBI:43451
IUPAC Name hydrogen iodide
SMILES I

Specifications

Boiling Point 127°C
Density 1.96g/mL at 20°C
Packaging Glass bottle
Quantity 50 mL
UN Number UN1787
Formula Weight 127.91g/mol
Percent Purity ≥67% (T)
Chemical Name or Material Hydriodic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'