missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles

Provides foot with side support and extra support at heel and middle of foot.

£31.30 - £34.35

Specifications

Color Gray
Product Type Stability Inlay Sole
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10556554
View Documents
Emma Safety Footwear
MM00107245
45
£34.35
1 pair
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
10129674
View Documents
Emma Safety Footwear
MM00107243
43
£31.30
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
10705092
View Documents
Emma Safety Footwear
MM00107246
46
£31.30
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
15173872
View Documents
Emma Safety Footwear
MM00107239
39
£31.30
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
10372410
View Documents
Emma Safety Footwear
MM00107241
41
£31.30
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
10357714
View Documents
Emma Safety Footwear
MM00107244
44
£31.30
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
Description

Description

  • Regulates moisture
  • Contains active carbon to prevent odor
Specifications

Specifications

Gray
Stability Inlay Sole
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.