missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid 44-49%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical

Product Code. 11974101 Shop All Fisher Chemical Products
Click to view available options
:
500mL
This item is not returnable. View return policy

Product Code. 11974101

Brand: Fisher Chemical undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH lt;1
Color Yellow
CAS 10035-10-6
Melting Point -11°C
Boiling Point 126°C
Vapor Pressure 8mm Hg 25
Quantity 500 mL
Molecular Formula BrH
Physical Form Liquid
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
SMILES Br
IUPAC Name hydrogen bromide
Molecular Weight (g/mol) 80.91
Formula Weight 80.9g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid 44-49%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.