missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 48%, Extra Pure, SLR, Fisher Chemical™
£90.65 - £303.00
Chemical Identifiers
CAS | 10035-10-6 |
---|---|
Molecular Formula | BrH |
Molecular Weight (g/mol) | 80.91 |
MDL Number | MFCD00011323 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-N |
Synonym | hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide |
PubChem CID | 260 |
ChEBI | CHEBI:47266 |
IUPAC Name | hydrogen bromide |
SMILES | Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10121860
|
Fisher Chemical
H/0900/PB08 |
500 mL |
£90.65
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10550341
|
Fisher Chemical
H/0900/PB17 |
2.5 L |
£303.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
10035-10-6 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
hydrogen bromide |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 | |
Br |
Specifications
lt;1 | |
10035-10-6 | |
126°C | |
8mm Hg 25 | |
Liquid | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
Br | |
80.91 | |
CHEBI:47266 |
Yellow | |
-11°C | |
HDPE plastic bottle | |
BrH | |
MFCD00011323 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
hydrogen bromide | |
260 | |
80.9g/mol |