missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, ≥99.99%; Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | Acidic |
Color | Yellow |
Composition | 48 wt.% in water |
Melting Point | -11°C |
Boiling Point | 126°C |
Density | 1.49g/mL at 25°C |
Odor | Stinging |
Packaging | Glass Bottle |
Vapor Pressure | 320psi (21.1°C), 8mmHg (25°C) |
Quantity | 100 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'