Learn More
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Melting Point | -11.0°C |
Boiling Point | 126.0°C to 128.0°C |
Density | 1.4800g/mL |
Heavy Metals (as Pb) | 5ppm max. |
Packaging | Glass bottle |
Specific Gravity | 1.48 |
Quantity | 500 mL |
Linear Formula | HBr |
Physical Form | Solution |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466153 | Certificate of Analysis | 25/09/2024 | 223320025,223325000 |
A0463084 | Certificate of Analysis | 23/05/2024 | 223320025,223325000 |
A0460154 | Certificate of Analysis | 15/02/2024 | 223320025,223325000 |
A0458445 | Certificate of Analysis | 07/12/2023 | 223320025,223325000 |
A0448153 | Certificate of Analysis | 30/11/2022 | 223320025,223325000 |
Safety and Handling


- Hydrobromic acid, ca. 48 wt% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.