missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid

Product Code. p-4527575 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
120 mL
480 mL
missing translation for 'unitSize'
Each
This item is not returnable. View return policy
Material is flammable before film is formed and HCl fumes are generated in its application. Fluid is acidic during application and care should be taken to avoid corrosion of metal that comes into contact with the liquid. After application, the surface is neutral.

Product Code. 11507590

missing translation for 'mfr': Thermo Scientific™ TS42800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Material is flammable before film is formed and HCl fumes are generated in its application. Fluid is acidic during application and care should be taken to avoid corrosion of metal that comes into contact with the liquid. After application, the surface is neutral.

For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid.

For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid. This short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface. It is excellent for modifying metals, certain plastics, ceramics and fiber optics.

Additional Product Information

Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.

The fluid is acidic only during application; after application, the surface is neutral.

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Corrosive to Eyes/Skin/Respiratory Tract
Standard Components 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane
Item Description 120 mL
Quantity 120 mL
Product Type Hydrocarbon Soluble Siliconizing Fluid
compliance-icons
Product Identifier
  • Surfasil, 120 mL.
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P501-Dispose of contents/container to .

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.