missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.01N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11303797
Click to view available options
Quantity:
1 L
4 L
20 L

Hydrochloric acid, 0.01N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Quantity 1 L
Color Colorless
IUPAC Name chlorane
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 0.083%; Water: 99.917%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water, methanol,ethanol and ether
Physical Form Liquid
Chemical Name or Material Hydrochloric acid, 0.01N

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'