Learn More
Hydrochloric acid, 0.1 N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124200250
Code : NEW
Additional Details : CAS Number : 7732-18-5 Weight : 25.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Hydrochloric acid | |
0.1 N Standard Solution | |
100.0°C | |
1.0000g/mL | |
1 | |
ClH | |
HCl | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 |
Colorless | |
7647-01-0,7732-18-5 | |
0.0998 to 0.1002N (20°C) | |
Plastic Can | |
25 L | |
Liquid | |
MFCD00011324 MFCD00792839 | |
15, 4818 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene | |
Cl | |
36.46 | |
CHEBI:17883 |
RUO – Research Use Only