missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1M, BAKER ANALYZED, J.T. Baker™

Product Code. 10269480
Click to view available options
Quantity:
10 L
Packaging:
PolyCube (HDPE)
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality grade volumetric solution is ready to use for laboratory and specialized industrial applications.

  • Standardized potentiometrically against an NIST Standard Reference Material, Sodium Carbonate
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

CAS 7647-01-0
Melting Point 0°C
Boiling Point 100°C
Concentration 0.0997 to 0.1003mol/L
Density 1g/cm3 at 20°C
Heavy Metals (as Pb) Max. 0.5 ppm
Packaging PolyCube (HDPE)
Normality 0.1N
Quantity 10 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Fully miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Grade Standardized potentiometrically against a NIST Standard Reference Material, Sodium Carbonate
Chemical Name or Material Hydrochloric acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'