Learn More
Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
£24.72 - £42.50
Chemical Identifiers
CAS | 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485030
|
Thermo Scientific Alfa Aesar
035611.AP |
500 mL |
£24.72
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393717
|
Thermo Scientific Alfa Aesar
035611.K2 |
1 L |
£32.36
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303727
|
Thermo Scientific Alfa Aesar
035611.K7 |
4 L | N/A | ||||||
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0, 7732-18-5 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
Colorless to pale yellow liquid | |
7647-01-0, 7732-18-5 | |
ClH | |
MFCD00011324 MFCD00792839 | |
Completely miscible with water. | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid: 10.0%; Water: 90.0% |
Colorless to Yellow | |
23 hPa (17mm Hg) at 20°C | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 | |
Hydrochloric acid |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
H290-H315-H318
missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: III
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Hydrochloric acid, 10% v/v aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.