missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124210025
43.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Hydrochloric acid | |
1N standard solution | |
0.998 to 1.002N (20°C) | |
Plastic Bottle | |
ClH | |
Liquid | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Colorless | |
7647-01-0, 7732-18-5 | |
1.0200g/mL | |
2.5 L | |
HCl | |
MFCD00011324 MFCD00792839 | |
15, 4818 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only