Learn More
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
£29.90 - £154.00
Specifications
Chemical Name or Material | Hydrochloric acid |
---|---|
Color | Colorless |
Name Note | 1N standard solution |
CAS | 7647-01-0, 7732-18-5 |
Concentration | 0.998 to 1.002N (20°C) |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10477700
|
Thermo Scientific Acros
124210010 |
1 L | Plastic Bottle |
£29.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703314
|
Thermo Scientific Acros
124210025 |
2.5 L | Plastic Bottle |
£54.90
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15214624
|
Thermo Scientific Acros
124210100 |
10 L | Plastic drum |
£154.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Hydrochloric acid | |
1N standard solution | |
0.998 to 1.002N (20°C) | |
ClH | |
Liquid | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
313 |
Colorless | |
7647-01-0, 7732-18-5 | |
1.0200g/mL | |
HCl | |
MFCD00011324 MFCD00792839 | |
15, 4818 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
36.46 | |
CHEBI:17883 |
Safety and Handling
GHS H Statement:
May be corrosive to metals.
GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Hydrochloric acid, 1N standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only