Learn More
Hydrochloric Acid 2 M According to European Pharmacopoeia (EP), Reagecon
Fully factorized, high-purity, stable product developed and tested for titrations.
Brand: Reagecon 3001700
Code : A0
Additional Details : Weight : 0.01000kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.2%.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.
- NIST traceable.
Specifications
Accuracy ± 0.2% | |
HDPE Bottle GL45 | |
Titration and EP Monographs |
2 M | |
1 L | |
Liquid |
Safety and Handling
- HYDROCHLORIC ACID 2M
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid