missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 32-35%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Product Code. p-7072848 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 11974081

Brand: Fisher Chemical A466250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Vapor Pressure 125mbar at 20°C
Quantity 250 mL
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
Formula Weight 36.46g/mol
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 32-35%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P353-Rinse skin with water/shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.