missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™ Product Code.: 10294190

Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™

Product Code. 10294190
1 L, HDPE plastic bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10294190

Brand: Fisher Chemical H/1200/PB15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, chlorwasserstoff, chloorwaterstof, muriatic acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883

Specifications

pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Packaging HDPE plastic bottle
Vapor Pressure 125mbar at 20°C
Quantity 1 L
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
MDL Number MFCD00011324
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
SMILES [H+].[Cl-]
IUPAC Name chlorane
Molecular Weight (g/mol) 36.458
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.